siRNA Details
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107024.1 | NP_001100494.1 | 2, 3 | 382 | ||
XM_006246905.2 | XP_006246967.1 | 3, 4 | 298 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107024.1 | NP_001100494.1 | 2, 3 | 382 | ||
XM_006246905.2 | XP_006246967.1 | 3, 4 | 298 |