siRNA Details
Gene Symbol: Atg9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191968.1 | NP_001178897.1 | 6 | 1062 | ||
XM_006235960.2 | XP_006236022.1 | 6 | 1062 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191968.1 | NP_001178897.1 | 6 | 1062 | ||
XM_006235960.2 | XP_006236022.1 | 6 | 1062 |