siRNA Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022189.1 | NP_071525.1 | 8 | 957 | ||
GenBank mRNA | AF155044.1 | - | 8 | 957 | ||
AF303447.1 | - | 624 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022189.1 | NP_071525.1 | 8 | 957 | ||
GenBank mRNA | AF155044.1 | - | 8 | 957 | ||
AF303447.1 | - | 624 |