siRNA Details
Gene Symbol: Kcnk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022293.1 | NP_071629.1 | 1 | 389 | ||
XM_006240447.2 | XP_006240509.1 | 2 | 3187 | |||
GenBank mRNA | AF287301.1 | - | 1 | 389 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022293.1 | NP_071629.1 | 1 | 389 | ||
XM_006240447.2 | XP_006240509.1 | 2 | 3187 | |||
GenBank mRNA | AF287301.1 | - | 1 | 389 |