siRNA Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 3, 4 | 342 | ||
GenBank mRNA | AF071225.1 | - | 3, 4 | 342 | ||
BC061971.1 | - | 3, 4 | 371 | 5623342 | ||
FQ211358.1 | - | 3, 4 | 346 | |||
FQ212031.1 | - | 3, 4 | 371 | |||
FQ219792.1 | - | 3, 4 | 380 | |||
FQ219926.1 | - | 3, 4 | 392 | |||
FQ221020.1 | - | 3, 4 | 395 | |||
FQ221248.1 | - | 3, 4 | 390 | |||
FQ222696.1 | - | 3, 4 | 504 | |||
FQ223162.1 | - | 3, 4 | 389 | |||
FQ229174.1 | - | 3, 4 | 392 | |||
FQ229214.1 | - | 3, 4 | 386 | |||
FQ230004.1 | - | 396 | ||||
FQ231656.1 | - | 3, 4 | 385 | |||
FQ232077.1 | - | 3, 4 | 381 | |||
FQ232920.1 | - | 3, 4 | 395 |
