siRNA Details
Gene Symbol: LOC101928514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110837.1 | 2 | 1261 | |||
GenBank mRNA | AK055500.1 | - | 2 | 1265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000585863.2 | - | 2 | 1261 | Chr. 17 - 76799044 - 76805953 [+] on Build GRCh38 |
