siRNA Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000855.2 | NP_000846.1 | 8 | 4277 | ||
| NM_001256424.1 | NP_001243353.1 | 9 | 4370 | |||
| GenBank mRNA | AK098142.1 | - | 1 | 929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY1A2| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000855.2 | NP_000846.1 | 8 | 4277 | ||
| NM_001256424.1 | NP_001243353.1 | 9 | 4370 | |||
| GenBank mRNA | AK098142.1 | - | 1 | 929 | ||