siRNA Details
| Gene Symbol: LACTB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038881.1 | 4 | 3549 | |||
| GenBank mRNA | AL833348.1 | - | 4 | 3548 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

LACTB2-AS1| Gene Symbol: LACTB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038881.1 | 4 | 3549 | |||
| GenBank mRNA | AL833348.1 | - | 4 | 3548 | ||
Genomic Map