siRNA Details
Gene Symbol: 4933431G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045163.1 | 3 | 809 | |||
NR_045165.1 | 3 | 1213 | ||||
GenBank mRNA | AK016225.1 | - | 3 | 812 | ||
AK017004.1 | - | 3 | 1214 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933431G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045163.1 | 3 | 809 | |||
NR_045165.1 | 3 | 1213 | ||||
GenBank mRNA | AK016225.1 | - | 3 | 812 | ||
AK017004.1 | - | 3 | 1214 |