siRNA Details
Gene Symbol: LOC101927869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109963.1 | 4 | 1106 | |||
GenBank mRNA | BC043533.1 | - | 4 | 1106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416218.1 | - | 4 | 1106 | Chr. 21 - 24304550 - 24321377 [-] on Build GRCh38 |
