siRNA Details
Gene Symbol: LOC105377621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136202.1 | 5 | 391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505709.1 | - | 2 | 233 | Chr. 4 - 102418602 - 102431282 [-] on Build GRCh38 |
