siRNA Details
Gene Symbol: LINC00593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026764.1 | 5 | 1311 | |||
GenBank mRNA | BC031958.1 | - | 5 | 1311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000560853.5 | - | 5 | 1311 | Chr. 15 - 69835234 - 69843120 [+] on Build GRCh38 |
