siRNA Details
| Gene Symbol: LOC102723640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135684.1 | 3 | 322 | |||
| GenBank mRNA | HY009132.1 | - | 3 | 323 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723640| Gene Symbol: LOC102723640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135684.1 | 3 | 322 | |||
| GenBank mRNA | HY009132.1 | - | 3 | 323 | ||
Genomic Map