siRNA Details
Gene Symbol: LINC00221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027457.2 | 3 | 782 | |||
GenBank mRNA | BX161407.1 | - | 2 | 585 | ||
BX248748.1 | - | 3 | 754 | |||
BX356743.2 | - | 3 | 708 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027457.2 | 3 | 782 | |||
GenBank mRNA | BX161407.1 | - | 2 | 585 | ||
BX248748.1 | - | 3 | 754 | |||
BX356743.2 | - | 3 | 708 |