siRNA Details
| Gene Symbol: Gm29508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076979.1 | - | 3 | 681 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB019254.2 | - | 3 | 618 | Chr. 12 - 103312368 - 103314708 [-] on Build GRCm38 | |

Gm29508
Genomic Map