siRNA Details
| Gene Symbol: Eef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017245.2 | NP_058941.1 | 10 | 2344 | ||
| GenBank mRNA | BC066661.1 | - | 10 | 2344 | 5623322 | |
| FQ215372.1 | - | 2 | 588 | |||
| FQ220112.1 | - | 2 | 580 | |||
| FQ229435.1 | - | 2 | 398 | |||

Eef2
Genomic Map