siRNA Details
| Gene Symbol: Ntf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270868.1 | NP_001257797.1 | 3 | 406 | ||
| NM_001270869.1 | NP_001257798.1 | 2 | 368 | |||
| NM_001270870.1 | NP_001257799.1 | 2 | 340 | |||
| NM_031073.3 | NP_112335.3 | 3 | 450 | |||
| XM_008763294.1 | XP_008761516.1 | 2 | 422 | |||
| XM_008763295.1 | XP_008761517.1 | 2 | 402 | |||
| GenBank mRNA | BC070504.1 | - | 3 | 450 | 7097740 | |
| EV778881.1 | - | 2 | 357 | |||
| FM074856.1 | - | 3 | 405 | |||
| FM101680.1 | - | 2 | 367 | |||
| M34643.1 | - | 2 | 201 | |||

Ntf3
Genomic Map