siRNA Details
| Gene Symbol: Reps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106264.1 | NP_001099734.1 | 1891 | |||
| Gene Symbol: LOC100909392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758664.1 | XP_008756886.1 | 5 | 950 | ||

Reps1
Genomic Map