siRNA Details
Gene Symbol: Mef2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014035.1 | NP_001014057.1 | 5 | 687 | ||
XM_008759540.1 | XP_008757762.1 | 4 | 1349 | |||
XM_008759542.1 | XP_008757764.1 | 5 | 1427 | |||
GenBank mRNA | BC081907.1 | - | 5 | 687 | 7111112 | |
DQ323505.1 | - | 3 | 386 | |||
GU646868.1 | - | 3 | 386 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: BC096598, AK153751, AK040227.
Gene Ontology Categories:
