siRNA Details
| Gene Symbol: Ns5atp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207607.1 | NP_997490.1 | 1 | 194 | ||
| GenBank mRNA | AY533136.1 | - | 1 | 194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ns5atp4| Gene Symbol: Ns5atp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207607.1 | NP_997490.1 | 1 | 194 | ||
| GenBank mRNA | AY533136.1 | - | 1 | 194 | ||