siRNA Details
| Gene Symbol: Nmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109149.1 | NP_001102619.1 | 2 | 315 | ||
| XM_006229445.2 | XP_006229507.1 | 2 | 989 | |||
| XM_008759554.1 | XP_008757776.1 | 2 | 989 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmb| Gene Symbol: Nmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109149.1 | NP_001102619.1 | 2 | 315 | ||
| XM_006229445.2 | XP_006229507.1 | 2 | 989 | |||
| XM_008759554.1 | XP_008757776.1 | 2 | 989 | |||