siRNA Details
| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130576.1 | NP_001124048.1 | 2 | 794 | ||
| XM_008765518.1 | XP_008763740.1 | 3 | 3429 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130576.1 | NP_001124048.1 | 2 | 794 | ||
| XM_008765518.1 | XP_008763740.1 | 3 | 3429 | |||
Fam83a
Genomic Map