siRNA Details
| Gene Symbol: Tst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009437.4 | NP_033463.1 | 1, 2 | 618 | ||
| GenBank mRNA | AK003953.1 | - | 1, 2 | 625 | ||
| BC005644.1 | - | 1, 2 | 642 | 3257034 | ||
| CT010210.1 | - | 1, 2 | 592 | |||
| U35741.1 | - | 1, 2 | 606 | |||

Tst
Genomic Map