siRNA Details
Gene Symbol: CAMK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323374.1 | NP_001310303.1 | 6 | 531 | ||
NM_001323375.1 | NP_001310304.1 | 6 | 563 | |||
NM_001323376.1 | NP_001310305.1 | 3 | 533 | |||
NM_001323377.1 | NP_001310306.1 | 4 | 291 | |||
NM_001744.5 | NP_001735.1 | 5 | 679 | |||
GenBank mRNA | AK311662.1 | - | 3 | 388 | ||
AK312840.1 | - | 5 | 534 | |||
BC016695.1 | - | 5 | 523 | |||
BC025687.1 | - | 5 | 521 | 5205478 | ||
BG721313.1 | - | 5 | 538 | |||
D30742.1 | - | 5 | 509 | |||
L17000.1 | - | 5 | 473 | |||
L24959.1 | - | 6 | 531 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
