siRNA Details
| Gene Symbol: ZCWPW2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040432.3 | NP_001035522.1 | 3 | 318 | ||
| NM_001324169.1 | NP_001311098.1 | 2 | 198 | |||
| NM_001324170.1 | NP_001311099.1 | 2 | 198 | |||
| NR_136708.1 | 2 | 198 | ||||
| XM_017005755.1 | XP_016861244.1 | 2 | 3993 | |||
| XM_017005756.1 | XP_016861245.1 | 2 | 3996 | |||
| XM_017005757.1 | XP_016861246.1 | 2 | 4001 | |||
| XM_017005758.1 | XP_016861247.1 | 2 | 3996 | |||
| XM_017005759.1 | XP_016861248.1 | 2 | 3998 | |||
| GenBank mRNA | BC094696.1 | - | 3 | 298 | 30529418 | |
| DB451817.1 | - | 3 | 318 | |||

ZCWPW2
Genomic Map