siRNA Details
| Gene Symbol: MS4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000139.4 | NP_000130.1 | 3 | 350 | ||
| XM_005273846.4 | XP_005273903.1 | 4 | 665 | |||
| XM_011544850.2 | XP_011543152.1 | 4 | 283 | |||
| XM_017017362.1 | XP_016872851.1 | 4 | 665 | |||
| GenBank mRNA | AK304487.1 | - | 3 | 350 | ||
| AK310142.1 | - | 3 | 350 | |||
| BC074800.2 | - | 3 | 279 | 30915525 | ||
| BC074843.2 | - | 3 | 279 | 30915222 | ||
| D10583.1 | - | 3 | 298 | |||
| DA972757.1 | - | 3 | 350 | |||
| X66187.1 | - | 1 | 38 | |||

MS4A2
Genomic Map