siRNA Details
| Gene Symbol: Tbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011537.3 | NP_035667.1 | 6 | 1298 | ||
| GenBank mRNA | AF140427.1 | - | 6 | 1063 | ||
| AK044843.1 | - | 2 | 277 | |||
| BC090639.1 | - | 6 | 1302 | 30548234 | ||
| U57330.1 | - | 4 | 459 | |||

Tbx5
Genomic Map