siRNA Details
| Gene Symbol: Fbxw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013908.4 | NP_038936.1 | 7, 8 | 1383 | ||
| GenBank mRNA | AF176520.1 | - | 7, 8 | 1344 | ||
| AK019715.2 | - | 7, 8 | 1384 | |||
| AK154911.1 | - | 6, 7 | 2322 | |||
| AK188841.1 | - | 1, 2 | 87 | |||
| BC010776.1 | - | 7, 8 | 1310 | 4211592 | ||

Fbxw5
Genomic Map