siRNA Details
| Gene Symbol: Abhd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081119.1 | NP_001074588.1 | 2 | 426 | ||
| NM_026868.1 | NP_081144.1 | 3 | 533 | |||
| GenBank mRNA | AK044407.1 | - | 2 | 280 | ||
| BC043690.1 | - | 2 | 239 | 5148187 | ||
| BY742957.1 | - | 2 | 427 | |||
| DV655120.1 | - | 3 | 286 | |||

Abhd13
Genomic Map