Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,2-Propanediol, 57-55-6, MFCD00064272, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 398039500ML
Encompass
Linear Formula: CH3CH(OH)CH2OH, Molecular Weight: 76.09, ACS reagent, ≥99.5%, Synonym: 1,2-PDO, Propylene glycol. 1,2-Propanediol may be used in the following processes: Green synthesis of propylene carbonate. Production of lactic acid by green catalytic oxidation. Propanal generation by catalytic dehydration.
Catalog No. NC0761134
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Sigma Aldrich Fine Chemicals Biosciences 1,2-Propanediol, 57-55-6, MFCD00064272, 500 mL
Your feedback has been submitted: Thank you for helping us improve our website.