Learn More
ACROBiosystems ACROBiosystems Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free
Supplier: ACROBiosystems CDGC52W6100UG
Recombinant Protein;100UG;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-C52W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.0 kDa (CD3E) and 14.4 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G contains no tag at the C-terminus.
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.