Learn More
ACROBiosystems Mouse FMC63 2.5mg
Supplier:  ACROBiosystems MBSC0082.5MG
Encompass_Preferred
ActiveMax Human Anti-FMC63 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human Anti-FMC63 Protein expressed from human 293 cells (HEK293).
Catalog No. 50-253-4462
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ACROBiosystems Mouse FMC63 2.5mg
Your feedback has been submitted: Thank you for helping us improve our website.