Learn More
Cayman Chemical ANTIBODY MOZ-IN-3 500ug
SDP
Supplier:  Cayman Chemical 27402500 UG
Encompass_Preferred
A dopamine D1 receptor antagonist (Ki 5 nM) selective for D1 over D2-4 receptors (Kis 3751 1000 and 5934 nM respectively) binds to D5 receptors (Ki 4 4 nM) inhibits food intake in a dose-dependent manner in rats (ED50 0 84 mg/kg) reduces ethanol intake in Sardinian alcohol-preferring rats and sucrose intake in water-deprived and water-sated rats without affecting food or total fluid intake suppresses cocaine-induced arrhythmias in anesthetized dogs
Catalog No. 50-293-9333
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cayman Chemical ANTIBODY MOZ-IN-3 500ug
Your feedback has been submitted: Thank you for helping us improve our website.