Learn More
Cayman Chemical Adenosine Dialdehyde, 34240-05-6, 10 mg
SDP
Supplier:  Cayman Chemical 1564410
Encompass_Preferred
Molecular Formula C10H11N5O4, Purity ≥95%, Formula Weight 265.2, Synonyms: 6-amino-α-(1-formyl-2-hydroxyethoxy)-9H-purine-9-acetaldehyde; An irreversible inhibitor of SAH hydrolase (IC50 = 40 nM); indirectly inhibits cellular methyltransferase activity.
Catalog No. NC1565024
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cayman Chemical Adenosine Dialdehyde, 34240-05-6, 10 mg
Your feedback has been submitted: Thank you for helping us improve our website.