Learn More
Bio X Cell InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Size: 5 MG
Supplier:  Bio X Cell BP00011R005MG
Encompass
InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Size: 5 MG, Isotype: Armenian hamster IgG, Applications: in vivo T cell depletion, in vitro T cell stimulation/activation, Immunofluorescence, Flow cytometry
Catalog No. 50-139-3504
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Bio X Cell InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Size: 5 MG
Your feedback has been submitted: Thank you for helping us improve our website.