Learn More
Bio X Cell InVivoPlus anti-mouse PD-1 (CD279), Clone: 29F.1A12, Size: 5 MG
Supplier:  Bio X Cell BP0273R005MG
Encompass
InVivoPlus anti-mouse PD-1 (CD279), Clone: 29F.1A12, Size: 5 MG, Isotype: rat IgG2a, kappa, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, Immunohistochemistry (frozen), Flow cytometry, Western blot
Catalog No. 50-139-3627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Bio X Cell InVivoPlus anti-mouse PD-1 (CD279), Clone: 29F.1A12, Size: 5 MG
Your feedback has been submitted: Thank you for helping us improve our website.