Learn More
AG Scientific Inc Aphidicolin, 5 MG
Supplier:  AG Scientific Inc A10265MG
Encompass_Preferred
Aphidicolin is a reversible inhibitor of nuclear DNA replication in eukaryotes and some animal viruses. It interferes with dCTP-specific binding sites on DNA polymerase-alpha. It also works on other DNA polymerases, however it does not work on DNA polymerase-alpha. Viruses inducing DNA polymerase-alpha, such as Herpes simplex, can be affected by aphidicolin because of its mechanism. It could be used for clinical study. Isolated from Nigrospora oryzae.CAS Number: 38966-21-1Molecular Weight: 338.5Chemical Formula: C20H34O4Solubility: Methanol: Clear colorless solution at 10 mg/mlStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6879
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AG Scientific Inc Aphidicolin, 5 MG
Your feedback has been submitted: Thank you for helping us improve our website.