Learn More
Aviva Systems Biology Corp C18orf25 Antibody - middle region (ARP51799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C18orf25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C18orf25. Alternate names: ARKL1, MGC12909, MGC87799ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60618
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Aviva Systems Biology Corp C18orf25 Antibody - middle region (ARP51799_P050) | 100uL
Your feedback has been submitted: Thank you for helping us improve our website.