Learn More
Immunology Consultants Affinity Purified Chicken anti-Green Fluorescent Protein Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants CGFP45AZ
Encompass_Preferred
Chickens were immunized with highly purified green-fluorescent protein (GFP) isolated directly from Aequorea victoria. Eggs were collected. Antibodies were immunoaffinity purified using the native GFP immobilized on a solid phase. This antibody reacts with GFP as determined by IEP and ELISA techniques. It is suitable for blotting and ELISA applications. This antibody may recognize variants of recombinant Aequorea green-fluorescent protein. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2090
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.