Learn More
ACROBiosystems Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;E.coli;100UG;GLX,GLRX,GRX,TTase-1,GLRX1,GLRX-1,GRX1,GRX-1
Supplier:  ACROBiosystems GLXH5149100UG
Encompass_Preferred
Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;100UG;Human GLRX1, His Tag (GLX-H5149) is expressed from E.coli cells. It contains AA Met 1 - Gln 106 (Accession # NP_001112362).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 12.7 kDa. The protein migrates as 12.7 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8683
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ACROBiosystems Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;E.coli;100UG;GLX,GLRX,GRX,TTase-1,GLRX1,GLRX-1,GRX1,GRX-1
Your feedback has been submitted: Thank you for helping us improve our website.