Learn More
Active Motif Histone H3K9me3 antibody (pAb)
SDP
Supplier:  Active Motif 39065
Encompass_Preferred
Active Motif's Histone H3K9me3 antibody (pAb) was raised in a Rabbit host. It has been validated for use in Chromatin Immunoprecipitation, Dot blot, Immunocytochemistry, Immunofluorescence and Western blot; it has been shown to react with Human samples, but it is predicted that it will react with a wide range of sample types. For more complete and up-to-date information, please visit the Active Motif website.
Catalog No. NC1847665
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Active Motif Histone H3K9me3 antibody (pAb)
Your feedback has been submitted: Thank you for helping us improve our website.