Learn More
ACROBiosystems ACROBiosystems Human CD47 Protein, Llama IgG2b Fc Tag, low endotoxin
Supplier: ACROBiosystems CD7H5251100UG
Recombinant Protein;100UG;Human CD47, Llama IgG2b Fc Tag, low endotoxin (CD7-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # Q08722-3).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 41.6 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, His Tag (Cat. No. SIA-H5225) at 0.5 μg/mL (100 μL/well) can bind Human CD47, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD7-H5251) with a linear range of 0.1-8 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.