Learn More
ACROBiosystems PE-Labeled Human CD79B, His Tag (CDB-HP2H6) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1).
Supplier:  ACROBiosystems CDBHP2H625TESTS
Encompass_Preferred
B-cell antigen receptor complex-associated protein beta chain (CD79b) is also known as B-cell-specific glycoprotein B29, Ig-beta,Immunoglobulin-associated B29 protein, B29 and IGB, which is a single-pass type I membrane protein containing one Ig-like V-type ( immunoglobulin-like ) domain and one ITAM domain.CD79b is required in cooperation with CD79A for initiation of the signal transduction cascade activated by the B-cell antigen receptor complex (BCR).CD79b can enhance phosphorylation of CD79A, possibly by recruiting kinases which phosphorylate CD79A or by recruiting proteins which bind to CD79A and protect it from dephosphorylation. Defects in CD79b are the cause of agammaglobulinemia type 6 (AGM6) that is a primary immunodeficiency characterized by profoundly low or absent serum antibodies and low or absent circulating B cells due to an early block of B-cell development.
Catalog No. 50-223-1531
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ACROBiosystems PE-Labeled Human CD79B, His Tag (CDB-HP2H6) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1).
Your feedback has been submitted: Thank you for helping us improve our website.