Learn More
ACROBiosystems ACROBiosystems Human LAIR1 / CD305 Protein, Fc Tag
Supplier:  ACROBiosystems LA1H5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LAIR-1, Fc Tag (LA1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human LAIR-1, Fc Tag (Cat. No. LA1-H5252) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-210-6691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ACROBiosystems ACROBiosystems Human LAIR1 / CD305 Protein, Fc Tag
Your feedback has been submitted: Thank you for helping us improve our website.