Learn More
ACROBiosystems Recombinant Protein;Cynomolgus LAG-3 / CD223 Protein, Mouse IgG2a Fc Tag;HEK293;100UG;LA3,LAG3,CD223,LAG-3,FDC
Supplier: ACROBiosystems LA3C52A0100UG
Recombinant Protein;Cynomolgus LAG-3 / CD223 Protein, Mouse IgG2a Fc Tag;100UG;Cynomolgus LAG-3, Mouse IgG2a Fc tag (LA3-C52A0) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LAG-3, Mouse IgG2a Fc tag (Cat. No. LA3-C52A0) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 Mab, Human IgG1 with a linear range of 20-78 ng/mL (QC tested).
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.