Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadence Science™ Malleable Stainless Steel Animal Feeding Needles
SDP

Designed to meet the special needs of pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals. Cadence Science™ Malleable Stainless Steel Animal Feeding Needles are sterile and disposable.

Supplier:  Cadence Science™ 9919

 View more versions of this product

Catalog No. 14-825-280

This item is not returnable. View return policy

Description

Description

  • Type 304 stainless steel construction
  • Malleable
  • Sterile
  • Disposable
  • 100/box, 20 x 5/sterile pouch
Specifications

Specifications

Animal Feeding Needle
Single-use
15G
3.7 in.
Stainless Steel (304)
Animal feeding
Sterile
2.9 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadence Science™ Malleable Stainless Steel Animal Feeding Needles > Gauge x L: 15G x 3.7 in.; Ball Dia.: 2.9mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.