Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld Mouse serum, sterile filtered, heat inactivated, 100 mL

Supplier:  Bioworld 306111462

Encompass_Preferred

Collected from 8-12 week-old sexually mature mice that are free of common parasites.Physiochemical constituents are measured to ensure lot-to-lot consistency.Sterility tested.Hemoglobin levels < 35 mg/dl.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.

Catalog No. 50-254-226


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Bioworld Mouse serum, sterile filtered, heat inactivated, 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.