Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid protein (P13L, R203K, G204R, G214C), His Tag
Supplier:  ACROBiosystems NUNC52HA100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa.
Catalog No. NC2010484
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid protein (P13L, R203K, G204R, G214C), His Tag
Your feedback has been submitted: Thank you for helping us improve our website.