Learn More
ACROBiosystems Recombinant Protein;Human R-Spondin 3 / RSPO3 (22-146) Protein;HEK293;50UG;RS6,RSPO3,RSPO-3,R-spondin 3,Rspondin 3,R-spondin-3,CRISTIN1,PWTSR,THSD2
Supplier: ACROBiosystems RS6H522250UG
Recombinant Protein;Human R-Spondin 3 / RSPO3 (22-146) Protein;50UG;Human R-Spondin 3, His Tag (RS6-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession # AAH22367).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 18 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.