Learn More
Enzo Life Sciences RUNX3 (human) monoclonal antibody (R3-6E9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX803318C100
Encompass_Preferred
Clone: R3-6E9. Isotype: IgG1. Immunogen: Recombinant human RUNX3 (runt-related transcription factor 3) (aa 191-300). Species reactivity: Human. Applications: IHC (PS), IP, WB. Formulation: Liquid. In PBS containing 10% glycerol and 0.02% sodium azide.
Catalog No. 50-200-9737
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Enzo Life Sciences RUNX3 (human) monoclonal antibody (R3-6E9) (100µg)
Your feedback has been submitted: Thank you for helping us improve our website.